missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-O-methyl-D-glucose, 99.1%, MP Biomedicals™
Click to view available options
Quantity:
1 g
10 g
Specifications
Color | White |
Quantity | 1 g |
Formula Weight | 194.2 |
Percent Purity | 0.991 |
Physical Form | Powder |
Chemical Name or Material | 3-O-methyl-D-glucose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'