Läs mer
3-O-Methyl-L-DOPA Monohydrate, TRC
Produktkod. 30007933
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007933 50 mg
30005740 25 mg
30007557 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007933 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30381550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 200630-46-2
Molekylformel C10 H13 N O4 . H2 O
Molekylvikt (g/mol) 229.23
Synonym L-Tyrosine, 3-methoxy-, monohydrate (9CI), 3-Methoxy-L-tyrosine monohydrate, 3-Methoxy-4-hydroxy-L-phenylalanine monohydrate, 3-O-Methyl-L-DOPA monohydrate, L-4-Hydroxy-3-methoxyphenylalanine monohydrate, (2S)-2-Amino-3-(4-hydroxy-3-methoxy-phenyl)propanoic acid hydrate (1:1)
IUPAC-namn (2S)-2-amino-3-(4-hydroxy-3-methoxyphenyl)propanoic acid;hydrate
LEDER O.COc1cc(C[C@H](N)C(=O)O)ccc1O
Specifikationer
Kemiskt namn eller material 3-O-Methyl-L-DOPA Monohydrate
InChI formel InChI=1S/C10H13NO4.H2O/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14;/h2-3,5,7,12H,4,11H2,1H3,(H,13,14);1H2/t7-;/m0./s1
Kvantitet 50 mg
Formel vikt 229.095