Läs mer
3'-O-Methyl Quercetin, TRC
Produktkod. 17952185
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17952185 1 mg
17903715 5 mg
17944675 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17952185 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3265751MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 480-19-3
Molekylformel C16 H12 O7
Molekylvikt (g/mol) 316.26
Synonym Isorhamnetin, Flavone, 3,4',5,7-tetrahydroxy-3'-methoxy- (8CI), Isorhamnetin (6CI), 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-1-benzopyran-4-one, 3,4',5,7-Tetrahydroxy-3'-methoxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)chromen-4-one, 3'-Methoxyquercetin, 3'-Methylquercetin, 3'-O-Methylquercetin, C.I. 75680, Isorhamnetol, Quercetin 3'-methyl ether
IUPAC-namn 3,5,7-trihydroxy-2-(4-hydroxy-3-methoxyphenyl)chromen-4-one
LEDER COc1cc(ccc1O)C2=C(O)C(=O)c3c(O)cc(O)cc3O2
Specifikationer
Kemiskt namn eller material Isorhamnetin
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C16H12O7/c1-22-11-4-7(2-3-9(11)18)16-15(21)14(20)13-10(19)5-8(17)6-12(13)23-16/h2-6,17-19,21H,1H3
Formel vikt 316.0583
Procent renhet >95%