Learn More
3-(Oct-2-en-1-yl)dihydrofuran-2,5-dione, 98%, BLD Pharm™
Product Code. 30980150
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30980150 1 kg 1kg
30979084 100 g 100g
30977063 25 g 25g
30980110 500 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30980150 Supplier BLD Pharm Supplier No. BD008232341000g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 42482-06-4
Molecular Formula C12H18O3
MDL Number MFCD00047133
Quantity 1 kg
SMILES CCCCC/C=C/CC1CC(OC1=O)=O
Molecular Weight (g/mol) 210.27
Percent Purity 98%
Chemical Name or Material 3-(Oct-2-en-1-yl)dihydrofuran-2,5-dione