missing translation for 'onlineSavingsMsg'
Läs mer

3-Oxo-1,2-benzoisothiazoline-2-acetic Acid Methyl Ester 1,1-Dioxide, TRC

Product Code. 30420045
Change view
Click to view available options
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30420045 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420045 Supplier Toronto Research Chemicals Supplier No. TRCO869775500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Methyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
CAS 6639-62-9
Molekylformel C10 H9 N O5 S
Kvantitet 500 mg
Synonym Methyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Piroxicam Imp. D (EP)
InChI formel InChI=1S/C10H9NO5S/c1-16-9(12)6-11-10(13)7-4-2-3-5-8(7)17(11,14)15/h2-5H,6H2,1H3
LEDER COC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC-namn methyl 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetate
Molekylvikt (g/mol) 255.25
Formel vikt 255.0201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.