Läs mer
3-Oxo-2-propylpentanoic Acid Methyl Ester, TRC
Produktkod. 30419453
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419453 1 g
30413367 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30419453 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8702801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391052-84-8
Molekylformel C9H16O3
Molekylvikt (g/mol) 172.22
Synonym 3-Ketovalproic Acid Methyl Ester
IUPAC-namn methyl 3-oxo-2-propylpentanoate
LEDER CCCC(C(=O)CC)C(=O)OC
Specifikationer
Kemiskt namn eller material 3-Oxo-2-propylpentanoic Acid Methyl Ester
Kvantitet 1 g
InChI formel InChI=1S/C9H16O3/c1-4-6-7(8(10)5-2)9(11)12-3/h7H,4-6H2,1-3H3
Formel vikt 172.1099