Läs mer
3-Oxo-4-aza-5Alpha-Alphandrost-1,5-diene-17Beta-carboxylic Acid Methyl Ester, TRC
Produktkod. 30409562
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30409562 10 mg
1 options
Produktkod. 30409562 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84727010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Oxo-4-aza-5alpha-alphandrost-1,5-diene-17beta-carboxylic Acid Methyl Ester
CAS 166896-62-4
Molekylformel C20H27NO3
Kvantitet 10 mg
Synonym [4aR-(4aalpha,4bbeta,6aalpha,7alpha,9abeta,9balpha)]-2,4a,4b,5,6,6a,7,8,9,9a,9b,10-Dodecahydro-4a,6a-dimethyl-3-oxo-1H-indeno[5,4-f]quinoline-7-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C20H27NO3/c1-19-10-8-14-12(13(19)5-6-15(19)18(23)24-3)4-7-16-20(14,2)11-9-17(22)21-16/h7,9,11-15H,4-6,8,10H2,1-3H3,(H,21,22)/t12-,13-,14-,15+,19-,20+/m0/s1
LEDER COC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn methyl (1S,3aS,3bS,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,6,9b,10,11-decahydroindeno[5,4-f]quinoline-1-carboxylate
Molekylvikt (g/mol) 329.43
Formel vikt 329.1991