Läs mer
3-Oxo-5beta-cholanoic Acid, TRC
Produktkod. 30409734
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30409734 500 mg
1 options
Produktkod. 30409734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO848490500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Oxo-5beta-cholanoic Acid
CAS 1553-56-6
Anteckningar om renhetsgrad HPLC
Molekylformel C24 H38 O3
Kvantitet 500 mg
Synonym 3-Oxo-5β-cholan-24-oic Acid, 3-Oxo- 5β-cholanic Acid, 3-Ketolithocholic Acid, 3-Oxo-5β-cholan-24-oic Acid, 3-Oxo-5β-cholanic acid, 3-Oxo-5β-cholanoic Acid, 3-Oxocholanic Acid, 3-Oxolithocholic Acid, 3-keto-5β-Cholan-24-oic Acid, 3-keto-5β-Cholanic Acid, 5β-Cholanic Acid-3-one, Dehydrolithocholic Acid, Dehydro-lithocholic Acid
InChI formel InChI=1S/C24H38O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-16,18-21H,4-14H2,1-3H3,(H,26,27)/t15-,16-,18+,19-,20+,21+,23+,24-/m1/s1
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (4R)-4-[(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvikt (g/mol) 374.56
Formel vikt 374.2821