Läs mer
3-Oxo-7Alpha,12Alpha-hydroxy-5Beta-cholanoic Acid, TRC
Produktkod. 30138422
Change view
Kvantitet:
250 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138422 2.5 g 2.5g
30135631 1 g -
30006127 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138422 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8568952.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2304-89-4
Molekylformel C23 H36 O6
Molekylvikt (g/mol) 408.53
Synonym (5β,7α,12α)-7,12-Hydroxy-3-oxo-cholan-24-oic Acid, 7α,12α-Dihydroxy-3-oxo-5β-cholan-24-oic Acid, 7α,12α-Dihydroxy-3-oxo-5β-cholanic Acid, 7α,12α-Dihydroxy-3-keto-5β-cholanoic Acid, 7α,12α-Dihydroxy-3-oxo-5β-chol-24-oic Acid, 7α,12α-Dihydroxy-3-oxo-5β-cholan-24-oic Acid, 7α,12α-Dihydroxy-3-oxo-5β-cholanic Acid, 7α,12α-Dihydroxy-3-oxo-5β-cholanoic Acid, 3-Oxo-7α,12α-dihydroxy-5β-cholanic Acid, 3-Oxo-7α,12α-dihydroxy-5β-cholanoic Acid, 3-Oxo-7α,12α-dihydroxycholan-24-oic Acid, 3-Oxodeoxycholic Acid, 5b-CHOLANIC ACID-7a,12a-DIOL-3-ONE
IUPAC-namn (4R)-4-[(5R,7R,8R,9S,10S,12S,13S,14S,17S)-7,12-dihydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-4-hydroxybutanoic acid
LEDER C[C@]12CCC(=O)C[C@H]1C[C@@H](O)[C@H]3[C@@H]4CC[C@H]([C@H](O)CCC(=O)O)[C@@]4(C)[C@@H](O)C[C@H]23
Specifikationer
Kemiskt namn eller material 3-Oxo-7alpha,12alpha-hydroxy-5beta-cholanoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C23H36O6/c1-22-8-7-13(24)9-12(22)10-18(26)21-15-4-3-14(17(25)5-6-20(28)29)23(15,2)19(27)11-16(21)22/h12,14-19,21,25-27H,3-11H2,1-2H3,(H,28,29)/t12-,14+,15-,16-,17+,18+,19-,21-,22-,23+/m0/s1
Formel vikt 408.2512
Procent renhet >95%