Läs mer
3-Oxo Ropinirole Hydrochloride, TRC
Produktkod. 30137693
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137693 50 mg
30137649 100 mg
17993705 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137693 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87040050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 221264-21-7
Molekylformel C16 H22 N2 O2 . Cl H
Molekylvikt (g/mol) 310.82
Synonym 1H-Indole-2,3-dione, 4-[2-(dipropylamino)ethyl]-, hydrochloride (1:1), 1H-Indole-2,3-dione, 4-[2-(dipropylamino)ethyl]-, monohydrochloride (9CI)
IUPAC-namn 4-[2-(dipropylamino)ethyl]-1H-indole-2,3-dione;hydrochloride
LEDER Cl.CCCN(CCC)CCc1cccc2NC(=O)C(=O)c12
Specifikationer
Kemiskt namn eller material 3-Oxo Ropinirole hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C16H22N2O2.ClH/c1-3-9-18(10-4-2)11-8-12-6-5-7-13-14(12)15(19)16(20)17-13;/h5-7H,3-4,8-11H2,1-2H3,(H,17,19,20);1H
Formel vikt 310.1448
Procent renhet >95%