Läs mer
3-Oxohexanedioic Acid, TRC
Produktkod. 17974855
Change view
Kvantitet:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974855 25 mg
17972375 250 mg
17940355 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17974855 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85682525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 689-31-6
Molekylformel C6 H8 O5
Molekylvikt (g/mol) 160.125
Synonym 3-Ketoadipic Acid, 3-Oxoadipic Acid, NSC 18511, β-Ketoadipic Acid, β-Oxoadipic Acid;
IUPAC-namn 3-oxohexanedioic acid
LEDER OC(=O)CCC(=O)CC(=O)O
Specifikationer
Kemiskt namn eller material 3-Oxohexanedioic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C6H8O5/c7-4(3-6(10)11)1-2-5(8)9/h1-3H2,(H,8,9)(H,10,11)
Formel vikt 160.0372