Läs mer
3-(p-Carboxyphenyl)propionic Acid Dimethyl Ester, TRC
Produktkod. 30350608
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30350608 1 g
1 options
Produktkod. 30350608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1780901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(p-Carboxyphenyl)propionic Acid Dimethyl Ester
CAS 40912-11-6
Molekylformel C12H14O4
Kvantitet 1 g
Synonym 4-(Methoxycarbonyl)benzenepropanoic Acid Methyl Ester, Methyl 4-(3-Methoxy-3-oxopropyl)benzoate
InChI formel InChI=1S/C12H14O4/c1-15-11(13)8-5-9-3-6-10(7-4-9)12(14)16-2/h3-4,6-7H,5,8H2,1-2H3
LEDER COC(=O)CCc1ccc(cc1)C(=O)OC
IUPAC-namn methyl 4-(3-methoxy-3-oxopropyl)benzoate
Molekylvikt (g/mol) 222.24
Formel vikt 222.0892