missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Chlorophenoxy)lactic Acid, TRC

Product Code. 30361328
Change view
Click to view available options
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30361328 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361328 Supplier Toronto Research Chemicals Supplier No. TRCC414865250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-(p-Chlorophenoxy)lactic Acid
CAS 7698-98-8
Molekylformel C9 H9 Cl O4
Kvantitet 250 mg
Synonym p-Chlorophenoxylactic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropanoic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropionic Acid, NSC 518713
InChI formel InChI=1S/C9H9ClO4/c10-6-1-3-7(4-2-6)14-5-8(11)9(12)13/h1-4,8,11H,5H2,(H,12,13)
LEDER OC(COc1ccc(Cl)cc1)C(=O)O
IUPAC-namn 3-(4-chlorophenoxy)-2-hydroxypropanoic acid
Molekylvikt (g/mol) 216.618
Formel vikt 216.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.