missing translation for 'onlineSavingsMsg'
Läs mer

3-(p-Chlorophenoxy)lactic Acid, TRC

Produktkod. 30351254
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30351254 500 mg
30366066 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351254 Supplier Toronto Research Chemicals Supplier No. TRCC414865500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-(p-Chlorophenoxy)lactic Acid
CAS 7698-98-8
Molekylformel C9 H9 Cl O4
Kvantitet 500 mg
Synonym p-Chlorophenoxylactic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropanoic Acid, 3-(4-Chlorophenoxy)-2-hydroxypropionic Acid, NSC 518713
InChI formel InChI=1S/C9H9ClO4/c10-6-1-3-7(4-2-6)14-5-8(11)9(12)13/h1-4,8,11H,5H2,(H,12,13)
LEDER OC(COc1ccc(Cl)cc1)C(=O)O
IUPAC-namn 3-(4-chlorophenoxy)-2-hydroxypropanoic acid
Molekylvikt (g/mol) 216.618
Formel vikt 216.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.