Läs mer
3-(p-tert-Butylphenyl)isobutylaldehyde, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Lilial |
| CAS | 80-54-6 |
| Molekylformel | C14 H20 O |
| Kvantitet | 500 mg |
| Synonym | Hydrocinnamaldehyde, p-tert-butyl-α-methyl- (6CI,7CI,8CI), 4-(1,1-Dimethylethyl)-α-methylbenzenepropanal, (±)-2-Methyl-3-(4-tert-butylphenyl)propanal, 2-(4-tert-Butylbenzyl)propanal, 2-Methyl-3-(4-tert-butylphenyl)propanal, 2-Methyl-3-(p-tert-butylphenyl)propanal, 2-[(4-tert-Butylphenyl)methyl]propanal, 3-(4-tert-Butylphenyl)-2-methylpropanal, 3-(p-tert-Butylphenyl)-2-methylpropionaldehyde, 3-(p-tert-Butylphenyl)isobutylaldehyde, 4-tert-Butyl-α-methylhydrocinnamic aldehyde, Lilestralis, Lilial, Lysmeral, Lysmeral Extra, NSC 22275, lilestral, p-tert-Butyl-α-methyldihydrocinnamaldehyde, p-tert-Butyl-α-methylhydrocinnamaldehyde, p-tert-Butyl-α-methylhydrocinnamic aldehyde, pt-bucinal, α-Methyl-p-tert-butylhydrocinnamaldehyde, β-Lilial |
| InChI formel | InChI=1S/C14H20O/c1-11(10-15)9-12-5-7-13(8-6-12)14(2,3)4/h5-8,10-11H,9H2,1-4H3 |
| LEDER | CC(Cc1ccc(cc1)C(C)(C)C)C=O |
| IUPAC-namn | 3-(4-tert-butylphenyl)-2-methylpropanal |
| Molekylvikt (g/mol) | 204.31 |
| Formel vikt | 204.1514 |