Läs mer
3-Phenoxy-1,2-propanediol(>90%), TRC
Produktkod. 30412669
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30412669 50 mg
1 options
Produktkod. 30412669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31890850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Phenoxy-1,2-propanediol
CAS 538-43-2
Molekylformel C9 H12 O3
Kvantitet 50 mg
Synonym 1,2-Propanediol','3-phenoxy-, 3-Phenoxy-1,2-propanediol, 1,2-Dihydroxy-3-phenoxypropane, 1-Phenoxy-2,3-propanediol, 1-Phenoxymethyl-1,2-ethanediol, 2,3-Dihydroxypropyl phenyl ether, 3-Phenoxy-1,2-propoanediol, Antodyne, Glycerol α-monophenyl ether, Glycerol α-phenyl ether, NSC 406489, Phenol glyceryl ether, Phenylglyceryl ether, α-Phenyl monoglyceryl ether
InChI formel InChI=1S/C9H12O3/c10-6-8(11)7-12-9-4-2-1-3-5-9/h1-5,8,10-11H,6-7H2
LEDER OCC(O)COc1ccccc1
IUPAC-namn 3-Phenoxy-1,2-propanediol
Molekylvikt (g/mol) 168.19
Formel vikt 168.0786