Läs mer
3-(Phenoxy-13C6)benzoic Acid, TRC
Produktkod. 30012145
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012145 1 mg
30010323 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012145 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2280121MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1793055-05-6
Molekylformel C713C6H10O3
Molekylvikt (g/mol) 220.17
Synonym 3-PBA-13C6, m-(Phenyloxy-13C6)benzoic Acid, m-Phenoxybenzoic Acid-13C6
IUPAC-namn 3-((1,2,3,4,5,6-13C6)cyclohexatrienyloxy)benzoic acid
LEDER O=C(O)C1=CC(O[13C]2=[13C]([H])[13C]([H])=[13C]([H])[13C]([H])=[13C]2[H])=CC=C1
Specifikationer
Kemiskt namn eller material 3-(Phenoxy-13C6)benzoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C13H10O3/c14-13(15)10-5-4-8-12(9-10)16-11-6-2-1-3-7-11/h1-9H,(H,14,15)/i1+1,2+1,3+1,6+1,7+1,11+1
Formel vikt 220.0831
Procent renhet >95%