Läs mer
3-phenyl-1H-pyrrole-2-carboxylic acid, TRC
Produktkod. 30428692
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428692 100 mg
30420362 10 mg
30427584 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428692 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP322343100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 802052-63-7
Molekylformel C11H9NO2
Molekylvikt (g/mol) 187.19
IUPAC-namn 3-phenyl-1H-pyrrole-2-carboxylic acid
LEDER OC(=O)c1[nH]ccc1c2ccccc2
Specifikationer
Kemiskt namn eller material 3-phenyl-1H-pyrrole-2-carboxylic acid
Kvantitet 100 mg
InChI formel InChI=1S/C11H9NO2/c13-11(14)10-9(6-7-12-10)8-4-2-1-3-5-8/h1-7,12H,(H,13,14)
Formel vikt 187.0633