Learn More
3-Phenyl-N,N-bis(3-phenylpropyl)propan-1-amine Hydrochloride, Mikromol™
Product Code. 30052059
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052059 25 mg 25mg
30050480 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052059 Supplier Mikromol Supplier No. MM0454.060025
 Restricted Item
Beräknad leverans: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Phenyl-N,N-bis(3-phenylpropyl)propan-1-amine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Phenyl-N,N-bis(3-phenylpropyl)propan-1-amine Hydrochloride
CAS 878784-75-9
Analyte or Component Names 3-Phenyl-N,N-bis(3-phenylpropyl)propan-1-amine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C27 H33 N . Cl H
Certifications/Compliance ISO 17025
Synonym Alverine Citrate Imp. E (EP), Alverine Imp. E (EP), 3-Phenyl-N,N-bis(3-phenylpropyl)propan-1-amine Hydrochloride, Alverine Citrate Imp. E (EP) as Hydrochloride, Alverine Citrate Impurity E as Hydrochloride, Alverine Impurity E as Hydrochloride
InChI Formula InChI=1S/C27H33N.ClH/c1-4-13-25(14-5-1)19-10-22-28(23-11-20-26-15-6-2-7-16-26)24-12-21-27-17-8-3-9-18-27;/h1-9,13-18H,10-12,19-24H2;1H
SMILES Cl.C(CN(CCCc1ccccc1)CCCc2ccccc2)Cc3ccccc3
IUPAC Name 3-phenyl-N,N-bis(3-phenylpropyl)propan-1-amine;hydrochloride
Molecular Weight (g/mol) 408.02
Quantity 25 mg
Formula Weight 407.238 g/mol