Läs mer
3-Phenylcyclohexanone, TRC
Produktkod. 30422039
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422039 500 mg
30411773 250 mg
30414643 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422039 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP400243500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20795-53-3
Molekylformel C12H14O
Molekylvikt (g/mol) 174.24
Synonym 3-Phenylcyclohexanone, (±)-3-Phenylcyclohexanone, NSC 119037
IUPAC-namn 3-phenylcyclohexan-1-one
LEDER O=C1CCCC(C1)c2ccccc2
Specifikationer
Kemiskt namn eller material 3-Phenylcyclohexanone
Kvantitet 500 mg
InChI formel InChI=1S/C12H14O/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-3,5-6,11H,4,7-9H2
Formel vikt 174.1045