missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylhydantoin, 95%, Thermo Scientific Chemicals

Catalog Number p-7054637
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2221-13-8
C9H8N2O2
176.18
MFCD00209486
RUEGAVIENIPHGK-UHFFFAOYSA-N
227930
3-phenylimidazolidine-2,4-dione
O=C1CNC(=O)N1C1=CC=CC=C1
This item is not returnable. View return policy
2221-13-8
C9H8N2O2
176.18
MFCD00209486
RUEGAVIENIPHGK-UHFFFAOYSA-N
227930
3-phenylimidazolidine-2,4-dione
O=C1CNC(=O)N1C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Synonym 3-phenyl-2,4-imidazolidinedione, 3-phenyl-imidazolidine-2,4-dione, n-phenyl-2,5-imidazolidione, 2,4-imidazolidinedione,3-phenyl, 3-phenyl-1,3-diazolidine-2,4-dione, 3-phenylhydantoine, 2,4-imidazolidinedione, 3-phenyl
Formula Weight 176.17
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Phenylhydantoin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'