Learn More
3-Phenylindole-2-carboxylic acid, 97%, Thermo Scientific™
Catalog Number p-7052437
Click to view available options
Quantity:
250 mg
1 g
6915-67-9
C15H11NO2
237.258
MFCD00974212
FEYRMXBCLPKSIJ-UHFFFAOYSA-N
3-phenylindole-2-carboxylic acid, phenylindolcarbonsaure, phenylindolecarboxylicacid, 1h-indole-2-carboxylic acid, 3-phenyl
2763657
3-phenyl-1H-indole-2-carboxylic acid
C1=CC=C(C=C1)C2=C(NC3=CC=CC=C32)C(=O)O
6915-67-9
C15H11NO2
237.258
MFCD00974212
FEYRMXBCLPKSIJ-UHFFFAOYSA-N
3-phenylindole-2-carboxylic acid, phenylindolcarbonsaure, phenylindolecarboxylicacid, 1h-indole-2-carboxylic acid, 3-phenyl
2763657
3-phenyl-1H-indole-2-carboxylic acid
C1=CC=C(C=C1)C2=C(NC3=CC=CC=C32)C(=O)O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Reactant for preparation of anticancer agents, derivatives of amino acids and peptides, primary acylureas.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 195°C to 197°C
Color White to Cream
Quantity 250 mg
Formula Weight 237.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Phenylindole-2-carboxylic acid