Läs mer
(3-Phenylpropyl)carbamic Acid Ethyl Ester, TRC
Produktkod. 30416726
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30416726 100 mg
1 options
Produktkod. 30416726 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP336255100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3-Phenylpropyl)carbamic Acid Ethyl Ester
CAS 185910-72-9
Molekylformel C12H17NO2
Kvantitet 100 mg
Synonym Ethyl N-(3-Phenylpropyl)carbamate, N-(3-Phenylpropyl)carbamic Acid Ethyl Ester
InChI formel InChI=1S/C12H17NO2/c1-2-15-12(14)13-10-6-9-11-7-4-3-5-8-11/h3-5,7-8H,2,6,9-10H2,1H3,(H,13,14)
LEDER CCOC(=O)NCCCc1ccccc1
IUPAC-namn ethyl N-(3-phenylpropyl)carbamate
Molekylvikt (g/mol) 207.27
Formel vikt 207.1259