Läs mer
(3-Phenylpropyl)carbamic Acid Ethyl Ester, TRC
Produktkod. 30427583
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30427583 1 g
1 options
Produktkod. 30427583 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3362551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3-Phenylpropyl)carbamic Acid Ethyl Ester
CAS 185910-72-9
Molekylformel C12H17NO2
Kvantitet 1 g
Synonym Ethyl N-(3-Phenylpropyl)carbamate, N-(3-Phenylpropyl)carbamic Acid Ethyl Ester
InChI formel InChI=1S/C12H17NO2/c1-2-15-12(14)13-10-6-9-11-7-4-3-5-8-11/h3-5,7-8H,2,6,9-10H2,1H3,(H,13,14)
LEDER CCOC(=O)NCCCc1ccccc1
IUPAC-namn ethyl N-(3-phenylpropyl)carbamate
Molekylvikt (g/mol) 207.27
Formel vikt 207.1259