missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Piperidinopyridine-2-carboxylic acid, 97%, Maybridge™
889.00 SEK - 1151.00 SEK
Chemical Identifiers
CAS | 898289-01-5 |
---|---|
Molecular Formula | C11H14N2O2 |
Molecular Weight (g/mol) | 206.245 |
MDL Number | MFCD09025831 |
InChI Key | QIWMAMIMWHCYGZ-UHFFFAOYSA-N |
Synonym | 3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl |
PubChem CID | 18525731 |
IUPAC Name | 3-piperidin-1-ylpyridine-2-carboxylic acid |
SMILES | C1CCN(CC1)C2=C(N=CC=C2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10163454
|
Maybridge
CC37601CB |
250mg | Amber Glass Bottle |
889.00 SEK
250mg |
09-02-2021 |
||||
10031004
|
Maybridge
CC37601DA |
1g | Amber Glass Bottle |
1151.00 SEK
1g |
09-02-2021 |
||||
Chemical Identifiers
898289-01-5 | |
206.245 | |
QIWMAMIMWHCYGZ-UHFFFAOYSA-N | |
18525731 | |
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O |
C11H14N2O2 | |
MFCD09025831 | |
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl | |
3-piperidin-1-ylpyridine-2-carboxylic acid |
Specifications
3-Piperidinopyridine-2-carboxylic acid | |
250mg | |
96.5 | |
97% | |
MFCD09025831 | |
QIWMAMIMWHCYGZ-UHFFFAOYSA-N | |
3-piperidin-1-ylpyridine-2-carboxylic acid | |
18525731 | |
97% |
Amber Glass Bottle | |
898289-01-5 | |
100.0 | |
C11H14N2O2 | |
3-piperidinopyridine-2-carboxylic acid, 3-piperidin-1-yl pyridine-2-carboxylic acid, 3-piperidin-1-yl picolinic acid, 3-piperidylpyridine-2-carboxylic acid, 2-pyridinecarboxylicacid, 3-1-piperidinyl | |
C1CCN(CC1)C2=C(N=CC=C2)C(=O)O | |
206.245 | |
206.24 |