Learn More
3-Pyrrolidin-1-ylbenzaldehyde, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
IUPAC Name | 3-pyrrolidin-1-ylbenzaldehyde |
Formula Weight | 175.23 |
Percent Purity | 95% |
Chemical Name or Material | 3-Pyrrolidin-1-ylbenzaldehyde |