Läs mer
3-(Salicyloylamino)-1,2,4-triazole, TRC
Produktkod. 30411764
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411764 500 mg
30411891 50 mg
30422759 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411764 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS884030500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 36411-52-6
Molekylformel C9H8N4O2
Molekylvikt (g/mol) 205.19
IUPAC-namn 2-hydroxy-N-(1,2λ^{4},4-triazacyclopenta-1,3-dien-3-yl)benzamide
LEDER Oc1ccccc1C(=O)NC2=NCN=[NH]2
Specifikationer
Kemiskt namn eller material 3-(Salicyloylamino)-1,2,4-triazole
Kvantitet 500 mg
InChI formel InChI=1S/C9H9N4O2/c14-7-4-2-1-3-6(7)8(15)12-9-10-5-11-13-9/h1-4,13-14H,5H2,(H,10,12,15)
Formel vikt 205.0726