Läs mer
3-sec-Amylphenyl N-Methylcarbamate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-sec-Amylphenyl N-Methylcarbamate |
| CAS | 2282-34-0 |
| Molekylformel | C13H19NO2 |
| Kvantitet | 250 mg |
| Synonym | 3-(1-Methylbutyl)phenol 1-(N-Methylcarbamate), Methylcarbamic Acid m-(1-Methylbutyl)phenyl Ester, Chevron RE 5353, ENT 27,127, OMS 227, Ortho 5,353, RE 5353, m-(1-Methylbutyl)phenyl Methylcarbamate, m-sec-Amylphenyl N-Methyl Carbamate |
| InChI formel | InChI=1S/C13H19NO2/c1-4-6-10(2)11-7-5-8-12(9-11)16-13(15)14-3/h5,7-10H,4,6H2,1-3H3,(H,14,15) |
| LEDER | CCCC(C)C1=CC(=CC=C1)OC(=O)NC |
| IUPAC-namn | (3-pentan-2-ylphenyl) N-methylcarbamate |
| Molekylvikt (g/mol) | 221.3 |
| Formel vikt | 221.1416 |