Läs mer
3-Sulfo-glycodeoxycholic Acid Disodium Salt, TRC
Produktkod. 30415180
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30415180 1 mg
30411117 5 mg
30414086 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415180 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS7892651MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66874-10-0
Molekylformel C26H41NNa2O8S
Molekylvikt (g/mol) 573.65
Synonym Cholane, Glycine Derivative, N-[(3α,5β,12α)-12-Hydroxy-24-oxo-3-(sulfooxy)cholan-24-yl]-glycine Disodium Salt
LEDER [H][C@@](CC1)([C@H](C)CCC(NCC(O[Na])=O)=O)[C@]2(C)[C@]1([H])[C@]3([H])CC[C@]4([H])C[C@H](OS(=O)(O[Na])=O)CC[C@]4(C)[C@@]3([H])C[C@@H]2O
Specifikationer
Kemiskt namn eller material 3-Sulfo-glycodeoxycholic Acid Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C26H43NO8S.2Na/c1-15(4-9-23(29)27-14-24(30)31)19-7-8-20-18-6-5-16-12-17(35-36(32,33)34)10-11-25(16,2)21(18)13-22(28)26(19,20)3;;/h15-22,28H,4-14H2,1-3H3,(H,27,29)(H,30,31)(H,32,33,34);;/q;2*+1/p-2/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-;;/m1../s1
Formel vikt 573.2348
Procent renhet >95%