Läs mer
3'-Sulphated Lewis X, Methyl Glycoside Sodium Salt (~90%), TRC
Produktkod. 30420292
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30420292 1 mg
1 options
Produktkod. 30420292 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6992501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3'-Sulphated Lewis X','Methyl Glycoside Sodium Salt
CAS 176166-23-7
Molekylformel C21H37NO18S
Kvantitet 1 mg
Synonym 3'-SO4-Gal1-b-4[FUC1-alpha-3]GlcNAc-b-O-Me','Methyl O-2-alpha-3-O-(3-sulfo-b-D-galactopyranosyl)-1-b-4-O-[(alpha-L-fucopyranosyl)-1-alpha-3]-2-alphacetamido-2-deoxy-b-D-glucopyranoside] Sodium Salt
InChI formel InChI=1S/C21H37NO18S/c1-6-11(26)13(28)14(29)20(35-6)39-17-10(22-7(2)25)19(34-3)37-9(5-24)16(17)38-21-15(30)18(41(32,33)40-31)12(27)8(4-23)36-21/h6,8-21,23-24,26-31H,4-5H2,1-3H3,(H,22,25)/t6-,8+,9+,10+,11+,12-,13+,14?,15+,16+,17+,18?,19+,20-,21-/m0/s1
LEDER CO[C@@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)C([C@H]2O)S(=O)(=O)OO)[C@H](O[C@@H]3O[C@@H](C)[C@@H](O)[C@@H](O)C3O)[C@H]1NC(=O)C
IUPAC-namn (2S,3S,5S,6R)-2-[(2R,3S,4R,5R,6R)-5-acetamido-2-(hydroxymethyl)-6-methoxy-4-[(2S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl]oxy-3,5-dihydroxy-6-(hydroxymethyl)oxane-4-sulfonoperoxoic acid
Molekylvikt (g/mol) 623.58
Formel vikt 623.1731