missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid, 98%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 218°C to 220°C |
Quantity | 250 mg |
Formula Weight | 239.05 |
Percent Purity | 98% |
Chemical Name or Material | 3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'