missing translation for 'onlineSavingsMsg'
Learn More

3-tert-Butylphenol, 99%, Thermo Scientific Chemicals

Catalog Number p-7027511
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
585-34-2
C10H14O
150.22
MFCD00002300
CYEKUDPFXBLGHH-UHFFFAOYSA-N
m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
11450
CHEBI:34348
3-tert-butylphenol
CC(C)(C)C1=CC=CC(O)=C1
This item is not returnable. View return policy
585-34-2
C10H14O
150.22
MFCD00002300
CYEKUDPFXBLGHH-UHFFFAOYSA-N
m-tert-butylphenol, phenol, 3-1,1-dimethylethyl, 3-t-butylphenol, 3-tert-butyl phenol, phenol, m-tert-butyl, 3-tert-butyl-phenol, 3-tertbutylphenol, 3-tert.butylphenol, m-tert.-butylphenol
11450
CHEBI:34348
3-tert-butylphenol
CC(C)(C)C1=CC=CC(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 46°C
Boiling Point 125°C to 130°C (20 mmHg)
Flash Point 108°C (228°F)
Quantity 5 g
UN Number UN2430
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material 3-tert-Butylphenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'