missing translation for 'onlineSavingsMsg'
Learn More

3-Thienylmethylamine, 97%, Thermo Scientific™

Catalog Number p-2700339
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
27757-86-4
C5H7NS
113.178
MFCD01529872
DUDAKCCDHRNMDJ-UHFFFAOYSA-N
3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
2776381
thiophen-3-ylmethanamine
C1=CSC=C1CN
This item is not returnable. View return policy
27757-86-4
C5H7NS
113.178
MFCD01529872
DUDAKCCDHRNMDJ-UHFFFAOYSA-N
3-thienylmethylamine, 3-aminomethyl thiophene, 3-thiophenemethanamine, 3-thiophenemethylamine, 1-thien-3-ylmethanamine, 3-thienylmethyl amine, 1-3-thienyl methanamine, thiophen-3-yl-methylamine, thiophen-3-yl methanamine, thien-3-yl methylamine
2776381
thiophen-3-ylmethanamine
C1=CSC=C1CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material 3-Thienylmethylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'