missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethy)pyridine-2-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4812869
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
87407-12-3
C7H4F3NO2
191.11
MFCD07375386
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl-2-picolinic acid, acmc-209qkb, ksc447o0j, 3-trifluoromethylpicolinic acid, trifluoromethyl-2-picolinic acid
13233844
3-(trifluoromethyl)pyridine-2-carboxylic acid
OC(=O)C1=C(C=CC=N1)C(F)(F)F
This item is not returnable. View return policy
87407-12-3
C7H4F3NO2
191.11
MFCD07375386
DMWLBOPLZYJGPT-UHFFFAOYSA-N
3-trifluoromethyl pyridine-2-carboxylic acid, 3-trifluoromethyl picolinic acid, 3-trifluoromethyl-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 3-trifluoromethyl, 3-trifluoromethyl-2-picolinic acid, acmc-209qkb, ksc447o0j, 3-trifluoromethylpicolinic acid, trifluoromethyl-2-picolinic acid
13233844
3-(trifluoromethyl)pyridine-2-carboxylic acid
OC(=O)C1=C(C=CC=N1)C(F)(F)F
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethy)pyridine-2-carboxylic acid
compliance-icons
Product Identifier
  • 3-(Trifluoromethy)pyridine-2-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'