missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)aniline, 99%, Thermo Scientific Chemicals

Catalog Number p-7022923
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
98-16-8
C7H6F3N
161.127
MFCD00007797
VIUDTWATMPPKEL-UHFFFAOYSA-N
3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
7375
3-(trifluoromethyl)aniline
C1=CC(=CC(=C1)N)C(F)(F)F
This item is not returnable. View return policy
98-16-8
C7H6F3N
161.127
MFCD00007797
VIUDTWATMPPKEL-UHFFFAOYSA-N
3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
7375
3-(trifluoromethyl)aniline
C1=CC(=CC(=C1)N)C(F)(F)F

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Solubility
Soluble in water. [5 g/L (20°C)]

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 5°C to 6°C
Density 1.295
Boiling Point 187°C to 189°C
Flash Point 85°C (185°F)
Odor Recognizable
Refractive Index 1.48
Quantity 100 g
UN Number UN2948
Beilstein 387672
Solubility Information Soluble in water. [5g/L (20°C)]
Formula Weight 161.13
Percent Purity 99%
Chemical Name or Material 3-(Trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'