Learn More
3-(Trifluoromethyl)benzaldehyde, TRC
Specifikationer
Specifikationer
| Chemical Name or Material | 3-(Trifluoromethyl)benzaldehyde |
| CAS | 454-89-7 |
| Molecular Formula | C8H5F3O |
| Quantity | 50 g |
| Synonym | 3-(Trifluoromethyl)phenylcarboxaldehyde, m-(Trifluoromethyl)benzaldehyde, α,α,α-Trifluoro-m-tolualdehyde |
| InChI Formula | InChI=1S/C8H5F3O/c9-8(10,11)7-3-1-2-6(4-7)5-12/h1-5H |
| SMILES | FC(F)(F)c1cccc(C=O)c1 |
| IUPAC Name | 3-(trifluoromethyl)benzaldehyde |
| Molecular Weight (g/mol) | 174.12 |
| Formula Weight | 174.0292 |