Learn More
3-(Trifluoromethyl)benzenepropanol, TRC
Product Code. 30457013
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457013 5 g 5g
30449749 100 mg 100mg
30456913 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30457013 Supplier Toronto Research Chemicals Supplier No. TRCT7911105G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78573-45-2
Molecular Formula C10H11F3O
Molecular Weight (g/mol) 204.19
Synonym 3-[3-(Trifluoromethyl)phenyl]propan-1-ol
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propan-1-ol
SMILES OCCCc1cccc(c1)C(F)(F)F
Specifications
Quantity 5 g
InChI Formula InChI=1S/C10H11F3O/c11-10(12,13)9-5-1-3-8(7-9)4-2-6-14/h1,3,5,7,14H,2,4,6H2
Formula Weight 204.0762
Chemical Name or Material 3-(Trifluoromethyl)benzenepropanol