Läs mer
3-Trifluoromethyl Hydroxychloroquine, TRC
Produktkod. 30449957
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449957 25 mg
30449256 2.5 mg
30451736 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30449957 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT81584025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2054858-88-5
Molekylformel C19H25ClF3N3O
Molekylvikt (g/mol) 403.87
Synonym 2-((4-((7-Chloro-3-(trifluoromethyl)quinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol
LEDER CCN(CCO)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1C(F)(F)F
Specifikationer
Kemiskt namn eller material 3-Trifluoromethyl Hydroxychloroquine
Kvantitet 25 mg
InChI formel InChI=1S/C19H25ClF3N3O/c1-3-26(9-10-27)8-4-5-13(2)25-18-15-7-6-14(20)11-17(15)24-12-16(18)19(21,22)23/h6-7,11-13,27H,3-5,8-10H2,1-2H3,(H,24,25)
Formel vikt 403.1638