Läs mer
3-Trimethylstannyl Benzoic Acid, TRC
Produktkod. 30456757
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456757 25 mg
30454159 10 mg
30454157 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30456757 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT79719025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161767-56-2
Molekylformel C10 H14 O2 Sn
Molekylvikt (g/mol) 284.93
IUPAC-namn 3-trimethylstannylbenzoic acid
LEDER C[Sn](C)(C)c1cccc(c1)C(=O)O
Specifikationer
Kemiskt namn eller material 3-Trimethylstannyl Benzoic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C7H5O2.3CH3.Sn/c8-7(9)6-4-2-1-3-5-6;;;;/h1-2,4-5H,(H,8,9);3*1H3;
Formel vikt 286.0016