Läs mer
3-Vinylpyridine (>90%), TRC
Produktkod. 30454581
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30454581 100 mg
30449970 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30454581 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV487500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1121-55-7
Molekylformel C7 H7 N
Molekylvikt (g/mol) 105.14
IUPAC-namn 3-ethenylpyridine
LEDER C=Cc1cccnc1
Specifikationer
Kemiskt namn eller material 3-Vinylpyridine
Kvantitet 100 mg
InChI formel InChI=1S/C7H7N/c1-2-7-4-3-5-8-6-7/h2-6H,1H2
Formel vikt 105.0578