Learn More
Masterflex™ 3-Way Large Bore Stopcock
Perfect for handling higher flow volumes in process while using connections similar to luer systems. Cole-Parmer™ 3-Way Large Bore Stopcock is non-sterile and not approved or intended for medical, clinical, surgical or other patient oriented applications.
Brand:  Masterflex™ 30600-23
Additional Details : Weight : 0.03600kg
Product Code. 11785069
  • 550.00 SEK / Pack of 10
Currently Unavailable: We have no information when this product will be available.
View alternative products
You may only order up to the available stock. For more details call customer services on +46 31 352 32 00.
Description
Description
  • Enhance fluid flow with a larger bore than standard luer stopcocks
  • Ideal for handling high flow volumes
Specifications
Specifications
3-way flow
Polycarbonate
10 Pack
Male Luer Lock, Female Luer