missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Almotriptan-d6 Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Almotriptan hydrochloride N,N-dimethyl-d6 |
| CAS | 1794782-57-2 |
| Molekylformel | C17 2H6 H19 N3 O2 S . Cl H |
| Kvantitet | 10 mg |
| Synonym | 2-[5-(Pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]-N,N-bis(trideuteriomethyl)ethanamine hydrochloride, Almotriptan-D6 hydrochloride, Almotriptan-D6 hydrochloride (N,N-dimethyl D6) |
| InChI formel | InChI=1S/C17H25N3O2S.ClH/c1-19(2)10-7-15-12-18-17-6-5-14(11-16(15)17)13-23(21,22)20-8-3-4-9-20;/h5-6,11-12,18H,3-4,7-10,13H2,1-2H3;1H/i1D3,2D3; |
| LEDER | Cl.[2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12)C([2H])([2H])[2H] |
| IUPAC-namn | 2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]-N,N-bis(trideuteriomethyl)ethanamine;hydrochloride |
| Molekylvikt (g/mol) | 377.96 |
| Formel vikt | 377.1811 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?