Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Brand:  RETSCH 02.407.0039
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10212501
  • 3265.00 SEK / Each
Estimated Shipment: 08-08-2024
to see stock.
Specifications
1Each
Ring Sieves
SR200/SR300 Rotor Beater Mills
8mm