Learn More
RETSCH 360° Stainless Steel Ring Sieve For SR200/SR300 Rotor Beater Mills
Brand:  RETSCH 02.407.0058
4203.75 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.15000kg
 View more versions of this product
Product Code. 10262401
  • 4425.00 SEK / Each
Estimated Shipment: 08-08-2024
to see stock.
Specifications
1Each
Ring Sieves
SR200/SR300 Rotor Beater Mills
1.25mm