missing translation for 'onlineSavingsMsg'
Läs mer

(3a,5b)-3-(Acetyloxy)pregnan-20-one, TRC

Produktkod. 30313059
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30313059 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30313059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA187895250MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material (3alpha,5beta)-3-(Acetyloxy)pregnan-20-one
CAS 1491-77-6
Molekylformel C23H36O3
Kvantitet 250 mg
Synonym 3alpha-Hydroxy-5beta-pregnan-20-one Acetate
InChI formel InChI=1S/C23H36O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h16-21H,5-13H2,1-4H3/t16-,17-,18+,19-,20+,21+,22+,23-/m1/s1
LEDER CC(=O)O[C@@H]1CC[C@@]2(C)[C@H](CC[C@H]3[C@@H]4CC[C@H](C(=O)C)[C@@]4(C)CC[C@H]23)C1
IUPAC-namn [(3R,5R,8R,9S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molekylvikt (g/mol) 360.53
Formel vikt 360.2664
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.