Läs mer
3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-methyl-(3aR,5R,6S,7R,7aR)-5H-pyrano[3,2-d]thiazole-6,7-diol, TRC
Produktkod. 30420931
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420931 5 mg
30415279 50 mg
30411937 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2936255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 179030-22-9
Molekylformel C8 H13 N O4 S
Molekylvikt (g/mol) 219.26
IUPAC-namn (5R,6S,7R)-5-(hydroxymethyl)-2-methyl-5,6,7,7a-tetrahydro-3aH-pyrano[3,2-d][1,3]thiazole-6,7-diol
LEDER CC1=NC2[C@@H](O)[C@H](O)[C@@H](CO)OC2S1
Specifikationer
Kemiskt namn eller material 3a,6,7,7a-Tetrahydro-5-(hydroxymethyl)-2-methyl-(3aR,5R,6S,7R,7aR)-5H-pyrano[3,2-d]thiazole-6,7-diol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C8H13NO4S/c1-3-9-5-7(12)6(11)4(2-10)13-8(5)14-3/h4-8,10-12H,2H2,1H3/t4-,5?,6-,7-,8?/m1/s1
Formel vikt 219.0565
Procent renhet >95%