Läs mer
(3a)-Allopregnanolone Sulfate Pyridinium Salt, TRC
Produktkod. 30137436
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137436 250 mg
30003447 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137436 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA547120250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1177357-94-6
Molekylformel C26H39NO5S
Molekylvikt (g/mol) 477.66
Synonym (3α,5α)-3-(Sulfooxy)pregnan-20-one Pyridinium Salt, 3α-Hydroxy-5α-pregnan-20-one Sulfate Pyridinium Salt;
IUPAC-namn [(3R,5S,8R,9S,10S,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] hydrogen sulfate;pyridine
LEDER CC(=O)[C@H]1CC[C@@H]2[C@@]1(CC[C@H]3[C@H]2CC[C@@H]4[C@@]3(CC[C@H](C4)OS(=O)(=O)O)C)C.C1=CC=NC=C1
Specifikationer
Kemiskt namn eller material (3alpha)-Allopregnanolone Sulfate Pyridinium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C21H34O5S.C5H5N/c1-13(22)17-6-7-18-16-5-4-14-12-15(26-27(23,24)25)8-10-20(14,2)19(16)9-11-21(17,18)3;1-2-4-6-5-3-1/h14-19H,4-12H2,1-3H3,(H,23,24,25);1-5H/t14-,15+,16-,17+,18-,19-,20-,21+;/m0./s1
Formel vikt 477.2549