Läs mer
(3Alpha,5Beta,6Alpha,7Alpha)-6,7-Epoxy-3-[(ethoxycarbonyl)oxy]cholan-24-oic Acid Methyl Ester, TRC
Produktkod. 30370878
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30370878 100 mg
1 options
Produktkod. 30370878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE891910100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3alpha,5beta,6alpha,7alpha)-6,7-Epoxy-3-[(ethoxycarbonyl)oxy]cholan-24-oic Acid Methyl Ester
CAS 125112-76-7
Molekylformel C28H44O6
Kvantitet 100 mg
Synonym 6,7-Epoxy-1H-cyclopenta[a]phenanthrene, Cholan-24-oic Acid Deriv.
InChI formel InChI=1S/C28H44O6/c1-6-32-26(30)33-17-11-13-28(4)20-12-14-27(3)18(16(2)7-10-22(29)31-5)8-9-19(27)23(20)25-24(34-25)21(28)15-17/h16-21,23-25H,6-15H2,1-5H3/t16-,17-,18-,19+,20+,21+,23+,24-,25+,27-,28-/m1/s1
LEDER CCOC(=O)O[C@@H]1CC[C@@]2(C)[C@@H](C1)[C@H]3O[C@H]3[C@H]4[C@@H]5CC[C@H]([C@H](C)CCC(=O)OC)[C@@]5(C)CC[C@H]24
Molekylvikt (g/mol) 476.65
Formel vikt 476.3138