Läs mer
3Alpha,7Beta-Di-O-acetyl Ursodeoxycholic Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3alpha,7beta-Di-O-acetyl Ursodeoxycholic Acid Methyl Ester |
| CAS | 60384-30-7 |
| Molekylformel | C29H46O6 |
| Kvantitet | 100 mg |
| Synonym | Methyl 3α,7β-Diacetoxy-5β-cholan-24-oate, Methyl Ursodeoxycholate Diacetate, 3α,7β-Dihydroxy-5β-cholanic Acid Methyl Ester Diacetate |
| InChI formel | InChI=1S/C29H46O6/c1-17(7-10-26(32)33-6)22-8-9-23-27-24(12-14-29(22,23)5)28(4)13-11-21(34-18(2)30)15-20(28)16-25(27)35-19(3)31/h17,20-25,27H,7-16H2,1-6H3/t17?,20-,21?,22+,23-,24-,25?,27-,28?,29?/m0/s1 |
| LEDER | [H][C@@]12[C@@H](OC(C)=O)C[C@]3([H])C[C@H](OC(C)=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CCC(OC)=O |
| IUPAC-namn | (3R,5S,7S,8R,9S,10S,13R,14S,17R)-17-((R)-5-methoxy-5-oxopentan-2-yl)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthrene-3,7-diyl diacetate |
| Molekylvikt (g/mol) | 490.67 |
| Formel vikt | 490.33 |