Läs mer
(3aR,4R,7S,7aS)-Octahydro-4,7-methano-1H-isoindole Hydrochloride, TRC
Produktkod. 30409661
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30409661 50 mg
1 options
Produktkod. 30409661 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23691550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (3aR,4R,7S,7aS)-Octahydro-4,7-methano-1H-isoindole Hydrochloride
CAS 1820572-35-7
Molekylformel C9H16N.(HCl)
Kvantitet 50 mg
Synonym (3aR,4R,7S,7aS)-Octahydro-1H-4,7-methanoisoindole Hydrochloride
InChI formel InChI=1S/C9H15N.ClH/c1-2-7-3-6(1)8-4-10-5-9(7)8;/h6-10H,1-5H2;1H/t6-,7+,8-,9+;
LEDER Cl.C1C[C@H]2C[C@@H]1[C@H]3CNC[C@@H]23
Molekylvikt (g/mol) 173.68
Formel vikt 173.0971